Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

BBL™ MacConkey II Agar, BD

Supplier:   BD
BBL™ MacConkey II Agar, BD
Supplier:  BD
Certificates
BBL™ MacConkey II Agar, BD
BBL™ MacConkey II Agar, BD

2 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

MacConkey II Agar is a selective and differential medium for the detection of coliform organisms and enteric pathogens.

  • Low concentration of bile salts
  • Crystal violet inhibits growth of gram-positive bacteria
  • Lactose and neutral red indicator aid in differentiation of enteric microorganisms

MacConkey II Agar is recommended for use with clinical specimens likely to contain mixed microbial flora, such as urine, because it allows a preliminary gropuing of enteric and other gram-negative bacteria.

Product Family Options

Product Information

  • Size
    Availability
    Price
  • MacConkey II agar
    500 g
    Log in for availability
    Log in for availability
    Each (500g)
    Each (500g) - $226.00
    $226.00
    Each (500g)
       $226.00

    Supplier Number: 212306
  • 90004-448
    MacConkey II agar
    MacConkey II agar
    10 kg
    Log in for availability
    Log in for availability
    Each
    Each - $2,990.00
    $2,990.00
    Each
       $2,990.00
     

    Supplier Number: 292861
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.