BBL™ Middlebrook 7H9 Broth with Glycerol, BD Life Sciences
90001-552
: BD
:
- Description:Middlebrook 7H9 Broth with Glycerol
- Size:5 mL
- Cat. No.:90001-552
- Supplier no.:221832
Middlebrook 7H9 Broth dehydrated base supplemented with glycerol supports the growth of mycobacteria, including M
- Inorganic salts provides essential growth substances
- Sodium citrate holds certain inorganic cations in solution
- Glycerol provides carbon and energy
- Malachite green dye partially inhibits bacterial contaminants
Middlebrook 7H9 Broth with appropriate supplements is primarily used for growth of pure cultures of mycobacteria for use in laboratory studies.
tuberculosis.