BBL™ Motility Test Medium, BD Life Sciences
Supplier: BD
Certificates
About this item
Motility Test Medium is a semisolid medium used for the detection of motility of enteric organisms.
- Growth spreads from line of inoculation if organism is motile
- Growth of nonmotile organisms occurs only along stab line
- Agar is the solidifying agent
Motility Test Medium is used for the detection of motility of gram-negative enteric bacilli.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.