ContactUsLinkComponent
Sanitizer, Wescodyne, PENETONE CORP
Sanitizer, Wescodyne, PENETONE CORP
  14003-552
Sanitizer, Wescodyne, PENETONE CORP
  14003-552
 :  751131
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Wescodyne Sanitizer, Dark Brown Liquid
  • Environmentally Preferable:
  • Cat. No.:
    14003-552
  • Supplier no.:
    751131
Wescodyne is an iodine-based sanitizer specially formulated as a hand and boot dip in the food processing industry.
  • High foaming at low concentration and when agitated
  • High level of tamed iodine
  • Mild on hands
  • Fast microbiocidal action and effective on most pathogenic microorganisms
  • Gives a brown coloration to its solution to indicate its presence. Product must be added when solution fades