Adapters, Pressure Vessel, Chemglass
Supplier: Chemglass
Certificates
About this item
Adapter permits rotary evaporation of the contents of any pressure vessels after the reaction is complete.
- Eliminates the need to transfer samples into a conventional round bottom flask
- Joint sizes other than 24/40 are available
- Bushing and o-ring included
Adapter can also be connected to any vacuum line having a 24/40 port or used in conjunction with a adapter to connect to hoses.
Delivery: Supplied complete with one open top bushing and Viton o-ring.