Alchohol Wipes, Techspray
14216-032
- Color:White
- Description:Alcohol, Isopropyl Wipes, Saturated
- Dimensions:5 1/4 x 51/2"
- Environmentally Preferable:
- Cat. No.:14216-032
- Supplier no.:1610-50PK
All-purpose cleaning wipes presaturated with pure, anhydrous isopropyl alcohol.
- Safe on plastics
- Non-corrosive
- Rapidly evaporating
- Zero residue
- Non-ozone depleting
Ideal for tape head cleaning, removal of fluxes, light oils, polar soils, and white mineral residue.
: EPA SNAP Approved.