ContactUsLinkComponent
Fast Flex™ Holding Solution, Epredia™
Fast Flex™ Holding Solution, Epredia™
Catalog # 89212-370
Supplier:  Epredia
CAS Number:  
Fast Flex™ Holding Solution, Epredia™
Catalog # 89212-370
Supplier:  Epredia
Supplier Number:  9800
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Fast Flex™ holding solution, 2.5 gal.
  • Cat. No.:
    89212-370
  • Supplier no.:
    9800
Specifications
About this item
Compatible with 10% Neutral Buffered Formalin and other fixatives
Cassettes to be palced in holding solution for at least 20 minutes prior to loading on the processor.