Order Entry
Puerto Rico
ContactUsLinkComponent
Defibrinated Sheep Blood, Bottles, Northeast Lab Services
Defibrinated Sheep Blood, Bottles, Northeast Lab Services
Catalog # 101374-414
CAS Number:  
Defibrinated Sheep Blood, Bottles, Northeast Lab Services
Catalog # 101374-414
Supplier Number:  SB100
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Media Format:
    Liquid
  • Size:
    4 oz.
  • Cat. No.:
    101374-414
  • Cap type:
    Crimp cap
  • Fill volume:
    100 ml
  • Supplier no.:
    SB100

Specifications

About this item

Four-ounce glass bottles contain Defibrinated Sheep Blood.

  • Glass Construction
  • Crimp Caps
  • Fill Volume: 100 mL
  • Bottle Size: Four Ounces

The bottles feature secure crimp caps.

NEL produces a wide variety of agar-based media in many sizes and types of bottles for clients in the pharmaceutical, hospital, food and beverage, cosmetic, and educational research industries.