ContactUsLinkComponent
CHROMagar™ KPC CHROMagar Base with KPC Supplement, Clinical Microbiology, Northeast Lab Services
CHROMagar™ KPC CHROMagar Base with KPC Supplement, Clinical Microbiology, Northeast Lab Services
Catalog # 102963-564
CAS Number:  
CHROMagar™ KPC CHROMagar Base with KPC Supplement, Clinical Microbiology, Northeast Lab Services
Catalog # 102963-564
Supplier Number:  CHRP0001
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Size:
    33.4 g
  • Cat. No.:
    102963-564
  • Supplier no.:
    CHRP0001
Specifications
About this item
CHROMagar Base is a unique culture medium for the direct isolation of bacteria harboring reduced susceptibility to carbapenems.
  • Detection after Overnight Incubation
  • Short Incubation
  • No Selective Pre-Enrichment Medium Needed
  • Direct Plating Option
A selective pre-enrichment medium is not needed, users can plate the sample directly, and incubation lasts only 18 to 24 hours.
The KPC Supplement has a shelf life of around two years for flexibility of use.
Caution: This item is for research use only.