Order Entry
Puerto Rico
ContactUsLinkComponent
Terminal Transferase 5X Buffer, Promega
Terminal Transferase 5X Buffer, Promega
Catalog # PAM1893
CAS Number:  
undefined
Terminal Transferase 5X Buffer, Promega
Catalog # PAM1893
Supplier Number:  M1893
CAS Number:  

Specifications

  • Size:
    3×500 µl
  • Storage Conditions:
    –30...–10 °C
  • Enzyme Name:
    Terminal Transferase 5X Buffer
  • Cat. No.:
    PAM1893
  • Supplier no.:
    M1893

Specifications

About this item

Terminal Transferase Buffer Pack