Order Entry
Puerto Rico
ContactUsLinkComponent
Ribonuclease H
Ribonuclease H
Certificates
undefined
Ribonuclease H
Certificates

Specifications

  • Storage Conditions:
    –30...–10 °C
  • Enzyme Name:
    Ribonuclease H
  • Concentration:
    0.5 - 2 units/µl
  • Tested Applications:
    Sequencing, cDNA Synthesis, cDNA Cloning

Specifications

Related Information