Order Entry
Puerto Rico
ContactUsLinkComponent
Recombinant Terminal Deoxynucleotidyl Transferase
Recombinant Terminal Deoxynucleotidyl Transferase
Certificates
undefined
Recombinant Terminal Deoxynucleotidyl Transferase
Certificates

Specifications

  • Enzyme type:
    Recombinant
  • Storage Conditions:
    –30...–10 °C
  • Enzyme Name:
    Terminal Deoxynucleotidyl Transferase
  • Concentration:
    30 units/µl
  • Tested Applications:
    Labeling in vitro, cDNA Cloning

Specifications

Related Information