Order Entry
Puerto Rico
ContactUsLinkComponent
M-MLV Reverse Transcriptase
M-MLV Reverse Transcriptase
Certificates
undefined
M-MLV Reverse Transcriptase
Certificates

Specifications

  • Storage Conditions:
    –30...–10 °C
  • Enzyme Name:
    M-MLV Reverse Transcriptase
  • Enzyme Synonyms:
    RT
  • Concentration:
    200 units/µl
  • Tested Applications:
    cDNA Synthesis, cDNA Cloning, Primer Extension

Specifications

Related Information