ICL Precision™ Dispenser with PTFE Tubing Kit, International Crystal Laboratories
Supplier: International Crystal Labs
Certificates
About this item
The Precision™ Dispenser with PTFE tubing has broad range of analytical applications of dispensing fluids with extreme accuracy better than 1%
- Feature low dead volume valveless piston action and synchronous motor drives
- PTFE surfaces eliminate the concern of cross contamination
- Capabile to function in a single or continuous mode, permitting the user to perform static or flow-through analysis
Three position Forward-Off-Reverse switch for control of flow direction, simply flush back the system with solvent in the reverse mode.
This can handle volumetric dispensing of aqueous solutions, solvents and acids.
Ordering information: The Precision™ Dispenser comes complete with a PTFE pump head and PTFE tubing kit. The connections system consisting of luer locks, PTFE tubing, nuts and Tefzel® ferrules can be ordered seperately.