Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Respirators
Respiratory Systems
Accessories for Positive Pressure Hood Systems, H-Series, 3M
Accessories for Positive Pressure Hood Systems, H-Series, 3M
43300-208
:
3M
Accessories for Positive Pressure Hood Systems, H-Series, 3M
43300-208
:
3M
:
7000130672
Color:
Metal
Description:
3M™ Thumbscrew Clamp W-5102-2
Cat. No.:
43300-208
For use with:
Whitecap™ Helmets
Supplier no.:
W-5102-2
Details:
—