Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Pentafluoropropionic anhydride for acylation derivatisation

Corrosive
Supplier:   Restek
Pentafluoropropionic anhydride for acylation derivatisation
Supplier:  Restek
Certificates
Pentafluoropropionic anhydride for acylation derivatisation

1 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Acylation derivatization reagent.

  • Most commonly used for electron capture detection
  • React with alcohols, amines, and phenols
  • Frequently used for drugs of abuse confirmation

Acylation reagents offer the same advantage available from silylation reagents: creating less polar, more volatile derivatives. In comparison to silylating reagents, the acylating reagents can more readily target highly polar multifunctional compounds, such as carbohydrates and amino acids. In addition, acylating reagents offer the distinct advantage of introducing electron-capturing groups, thus enhancing detectability during analysis.

Synonyms:  Perfluoropropanoic anhydride, Pentafluoropropanoic anhydride, PFPA
  • CAS Number:356-42-3
  • Formula:C₆F₁₀O₃
  • MDL Number:MFCD00000429
  • Molecular Weight:310.05
  • ADR:8,II
  •  :Density: 1.571 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • UN:3265
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Vial
    1 g
    Discontinued
    Supplier Number: -
  • Vial
    25 g
    Log in for availability
    Log in for availability
    Each
    Each - $155.01
    $155.01
    Each
       $155.01

    Supplier Number: 35621

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.