Micro Check Chlorine Test Strips
Catalog # 82013-210
Supplier: HF Scientific
CAS Number:
Specifications
- Description:Micro Check Test Strips, Chlorine
- Cat. No.:82013-210
- Supplier no.:09940
- No. of tests:50
Specifications
About this item
The Micro Check Test Strips are an quick, inexpensive and safe alternative for reagent chlorine testing.
- Quick and easy to use
- Available in free or total chlorine with sensitivity as low as 0.05 ppm