

Ethyl acetate ≥99.9%, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
:141-78-6
:
:CH₃COOC₂H₅
:MFCD00009171
:88.11 g/mol
:77 °C (1.013 hPa)
:–84 °C
:0.901 g/cm³ (lit.)
:11.5 %(V)
:Ambient
:205-500-4
:1173
:3,II
:GHS07
:GHS02
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental Certification:Chem21 Solvent selection guide recommended by default and after discussion
- Autoignition Temperature:801 °F
- Assay (GC):Min. 99.9 %
- Non-volatile matter:Max. 0.0005 %
- Water (Karl Fischer):Max. 0.02 %
- Suitability for residue analysis:Max. 5 ng/l
- Vapor Density:3 (20 °C, vs air)
- Vapor Pressure:73 mmHg ( 20 °C)
Frequently Bought Together