


Cyclohexane ≥99.5% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:110-82-7
:
Formula:C₆H₁₂
MDL Number:MFCD00003814
:84.16 g/mol
Boiling Pt:81 °C (1.013 hPa)
Melting Pt:6.4 °C
Density:0.78 g/cm³ (20 °C) (lit.)
Flash Pt:8.3 %(V)
Storage Temperature:Ambient
EINECS:203-806-2
UN:1145
ADR:3,II
:GHS07
:GHS08
:GHS02
Specification Test Results
- Assay (GC):Min. 99.5 %
- Non-volatile matter:Max. 0.001 %
- Water (Karl Fischer):Max. 0.01 %
- Suitability for residue analysis:Max. 5 ng/l
- Vapor Density:2.9 (vs air)
- Vapor Pressure:168.8 mmHg ( 37.7 °C)
- Explosion Limit:9 %
- Autoignition Temperature:500 °F