Order Entry
Puerto Rico
ContactUsLinkComponent
2-Propanol ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Flammable
Irritant
2-Propanol ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
   Sustainable Options Available
Certificates
undefined
Flammable
Irritant
2-Propanol ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental Certification:
    EPA safer chemical solvent (Green circle rating); Chem21 solvent selection guide recommended by default and after discussion

Specifications

Specification Test Results

  • IR spectrum:
    complying
  • Assay (GC):
    Min. 99.85 %
  • Non-volatile matter:
    Max. 0.0001 %
  • Water (Karl Fischer):
    Max. 0.02 %
  • Absorbance at 400 nm:
    Max. 0.005
  • Absorbance at 275 nm:
    Max. 0.005
  • Absorbance at 250 nm:
    Max. 0.30
  • Absorbance at 245 nm:
    Max. 1.0
  • Fluorescence (chinin) at 365 nm:
    Max. 0.5 ppb
  • GC (as heptachlor epoxide):
    Max. 10 ng/l
  • Vapor Density:
    2.1 (vs air)
  • Autoignition Temperature:
    750 °F
  • Explosion Limit:
    2 %
  • Vapor Pressure:
    33 mmHg ( 20 °C)

Specification Test Results