Order Entry
Puerto Rico
ContactUsLinkComponent
 
Dichloromethane ≥99.8% stabilized, CHROMASOLV™ for HPLC, Riedel-de Haën™
 
undefined
Dichloromethane ≥99.8% stabilized, CHROMASOLV™ for HPLC, Riedel-de Haën™
 

 

  • Appearance:
    complying
  • Identity (IR):
    complying
  • Assay (GC):
    Min. 99.80 %
  • Non-volatile matter:
    Max. 0.001 %
  • Water (Karl Fischer):
    Max. 0.02 %
  • Free acid (as HCl):
    Max. 0.001 %
  • Free chlorine:
    complying
  • Absorbance at 260 nm:
    Max. 0.01
  • Absorbance at 245 nm:
    Max. 0.10
  • Absorbance at 240 nm:
    Max. 0.30
  • Absorbance at 235 nm:
    Max. 0.70
  • Absorbance:
    complying
  • APHA:
    Max. 10
  • 2-methyl-2-buten (GC):
    10 - 60 ppm
  • Autoignition Temperature:
    1223 °F
  • Contains:
    amylene as stabilizer
  • Refractive Index:
    n20/D 1.424 (lit.)