Order Entry
Puerto Rico
ContactUsLinkComponent
Simmons Citrate Slant, Tubes, Northeast Lab Services
Simmons Citrate Slant, Tubes, Northeast Lab Services
Catalog # 101374-800
CAS Number:  
Simmons Citrate Slant, Tubes, Northeast Lab Services
Catalog # 101374-800
Supplier Number:  T1730
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Media Format:
    Liquid
  • Size:
    13 x 100 mm
  • Cat. No.:
    101374-800
  • Cap type:
    Screw cap
  • Fill volume:
    4 ml
  • Supplier no.:
    T1730

Specifications

About this item

Simmons tubes have a sturdy screw cap.

  • Screw Caps
  • Tubed Agar Serves a Variety of Lab Functions
  • Glass Tubes
  • Fill Volume: 3 mL
  • Size: 13 mm x 100 mm

From simple fermentation tubes to highly specialized clinical diagnostic media, NEL produces media tubes for a wide variety of clients including private laboratories, food processors, clinics and hospitals, academic institutions, and government agencies.