Order Entry
Puerto Rico
ContactUsLinkComponent
Nutrient Gelatin, Tubes, Northeast Lab Services
Nutrient Gelatin, Tubes, Northeast Lab Services
Catalog # 101374-738
CAS Number:  
Nutrient Gelatin, Tubes, Northeast Lab Services
Catalog # 101374-738
Supplier Number:  T1656
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Media Format:
    Liquid
  • Size:
    16 x 125 mm
  • Cat. No.:
    101374-738
  • Cap type:
    Screw cap
  • Fill volume:
    8 ml
  • Supplier no.:
    T1656

Specifications

About this item

Nutrient Gelatin tubes have a sturdy screw cap.

  • Screw Caps
  • Tubed Agar Serves a Variety of Lab Functions
  • Glass Tubes
  • Fill Volume: 8 mL

From simple fermentation tubes to highly specialized clinical diagnostic media, NEL produces media tubes for a wide variety of clients including private laboratories, food processors, clinics and hospitals, academic institutions, and government agencies.