To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Media Format:Liquid
- Size:4 oz.
- Cat. No.:101373-778
- Cap type:Aluminum crimp cap
- Fill volume:100 ml
- Supplier no.:B8110
Specifications
About this item
Fluid A includes a serum bottle, a rubber stopper, and an aluminum crimp cap.
- Rubber Stopper
- Aluminum Crimp Cap
- Fill Volume: 100 mL
The serum bottle is 4 oz., and Fluid A has a fill volume of 100 mL.
NEL manufactures over 500 proprietary products and over 1300 classic media formulations for clients in the pharmaceutical, hospital, food and beverage, cosmetic, and educational research industries.