Qualatherm® 73G 10" Qualatherm 450 Mid-Temperature Mitt, QRP
32889-011
: PIP
- Glove color:White
- Glove size:Large
- Ambidextrous/Hand-specific:Ambidextrous
- Disposable/Reusable:Reusable
- Powdered/Powder-free:Powder-Free
- Length:25.4 cm (10")
- Thickness:116 mil.
- Cat. No.:32889-011
- Supplier no.:73G
The QRP 73G mitt is a RoHS compliant mitten-style made of a PolyTuff silicon elastomer formulation that offers economical, clean, liquid-proof mid-temperature protection for wet or dry environments.
- Packaged for ISO 5 (Class 100)
- Wet or dry contact temperatures from –78 to 450 °F (–61.4 to 232 °C)
- Latex-Free
- Non-particulating, cut and abrasion resistant
- PolyTuff silicone elastomer is sulfur-free
- Excellent resistance to chemicals, solder and fluxes
Applications include: Microwave handling, autoclave handling, food processing.
Free from PCB, asbestos and fiberglass hazards; Micro-coated, high density, continuous filament nylon liner assures clean, easy donning and removal.