Order Entry
Puerto Rico
ContactUsLinkComponent
Todd Hewitt Broth, Tubes, Northeast Lab Services
Todd Hewitt Broth, Tubes, Northeast Lab Services
Catalog # 101374-866
CAS Number:  
Todd Hewitt Broth, Tubes, Northeast Lab Services
Catalog # 101374-866
Supplier Number:  T1865
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Media Format:
    Liquid
  • Description:
    Todd Hewitt Broth with CNA (LIM Broth)
  • Size:
    13 mm x 100 mm
  • Cat. No.:
    101374-866
  • Cap type:
    Screw cap
  • Fill volume:
    3 ml
  • Supplier no.:
    T1865

Specifications

About this item

Todd Hewitt Broth features CNA (LIM broth).

  • Prepared Medium
  • Polycarbonate Construction
  • Screw Cap
  • Fill Volume: 3 mL
  • CNA Featured

Tubes are constructed of polycarbonate.

NEL produces broth and other media tubes for a wide variety of clients including private laboratories, food processors, clinics and hospitals, academic institutions, and government agencies.