Order Entry
Puerto Rico
ContactUsLinkComponent
MacConkey Broth, Tubes, Northeast Lab Services
MacConkey Broth, Tubes, Northeast Lab Services
Catalog # 101374-618
CAS Number:  
MacConkey Broth, Tubes, Northeast Lab Services
Catalog # 101374-618
Supplier Number:  T1521
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Media Format:
    Liquid
  • Description:
    MacConkey Broth
  • Size:
    13 mm x 100 mm
  • Cat. No.:
    101374-618
  • Cap type:
    Screw cap
  • Fill volume:
    5 ml
  • Supplier no.:
    T1521

Specifications

About this item

MacConkey Broth is contained in a durable polycarbonate tube.

  • Prepared Medium
  • Polycarbonate Construction
  • Screw Cap
  • Fill Volume: 5 mL
  • Size: 13 mm x 100 mm

The tubes measure 13 mm x 100 mm and have secure screw caps.

NEL produces media tubes for a variety of clients including private laboratories, food processors, clinics and hospitals, academic institutions, and government agencies.