Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Media Format:Liquid
- Description:Brucella Agar Pour
- Size:16 mm x 125 mm
- Cat. No.:101374-532
- Cap type:Screw cap
- Fill volume:14 ml
- Supplier no.:T1186
Specifications
About this item
Brucella Agar Slant is a prepared medium contained in a 16 mm x 125 mm tube.
- Glass Construction
- Screw Cap
- Size: 16 mm x 125 mm
- Fill Volume: 14 mL
The slant has a fill volume of 14 mL.
NEL produces media tubes for a wide variety of clients including private laboratories, food processors, clinics and hospitals, academic institutions, and government agencies.