Order Entry
Puerto Rico
ContactUsLinkComponent
Anaerobic Laked Agar, Plates, Northeast Lab Services
Anaerobic Laked Agar, Plates, Northeast Lab Services
Catalog # 101374-336
CAS Number:  
Anaerobic Laked Agar, Plates, Northeast Lab Services
Catalog # 101374-336
Supplier Number:  P5060
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Media Format:
    Liquid
  • Description:
    Anaerobic Laked Agar with Kanamycin and Vancomycin
  • Size:
    15 mm x 100 mm
  • Cat. No.:
    101374-336
  • Fill volume:
    20 ml
  • Supplier no.:
    P5060

Specifications

About this item

Anaerobic Laked Agar contains Kanamycin and Vancomycin.

  • Fill Volume: 20 mL
  • Vancomycin Included
  • Kanamycin Included
  • Standard Petri Dish
  • Size: 15 mm x 100 mm

It's contained by a 15 mm x 100 mm standard petri dish.

The medium has a fill volume of 20 mL.

Certifications: ISO Class 5 / FS209E Class 100 Validated EC Rooms certified.