Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Media Format:Liquid
- Description:Violet Red Bile Glucose Agar
- Size:15 mm x 100 mm
- Cat. No.:101374-164
- Fill volume:20 ml
- Supplier no.:P1672
Chocolate Agar is contained in a standard petri dish.
- Fill Volume: 20 mL
- Prepared Medium
- Standard Petri Dish
- Size: 15 mm x 100 mm
The medium has a fill volume of 20 mL.
NEL's plated media operation utilizes carefully selected and validated ingredients for contaminant-free products.