Order Entry
Puerto Rico
ContactUsLinkComponent
Standard Methods Agar, Plates, Northeast Lab Services
Standard Methods Agar, Plates, Northeast Lab Services
Catalog # 101374-132
CAS Number:  
undefined
Standard Methods Agar, Plates, Northeast Lab Services
Catalog # 101374-132
Supplier Number:  P1656
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Media Format:
    Liquid
  • Description:
    Standard Methods Agar (Plate Count Agar)
  • Size:
    15 mm x 100 mm
  • Cat. No.:
    101374-132
  • Fill volume:
    20 ml
  • Supplier no.:
    P1656

Specifications

About this item

Standard Methods Agar is a prepared medium with a fill volume of 20 mL.

  • Fill Volume: 20 mL
  • Plate Count Agar
  • Prepared Medium
  • Standard Petri Dish
  • Size: 15 mm x 100 mm

It's contained in a standard petri dish measuring 15 mm x 100 mm.

NEL manufactures over 500 proprietary products and over 1300 classic media formulations for clients in the pharmaceutical, hospital, food and beverage, cosmetic, and educational research industries.

Certifications: ISO Class 5 / FS209E Class 100 Validated EC Rooms certified.