Order Entry
United States
ContactUsLinkComponent
Horse Blood Overlay Medium, Plates, Northeast Lab Services
Horse Blood Overlay Medium, Plates, Northeast Lab Services
  101373-986
 :  
Horse Blood Overlay Medium, Plates, Northeast Lab Services
  101373-986
 :  P1315
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Media Format:
    Liquid
  • Description:
    Horse Blood Overlay Medium (Bottom Layer with Columbia BA Base, Top Layer with Columbia BA Base undefoliated Horse Blood), Plated
  • Size:
    15 mm x 100 mm
  • Environmentally Preferable:
  • Cat. No.:
    101373-986
  • Fill volume:
    20 ml
  • Supplier no.:
    P1315

 

 

Horse Blood Overlay Medium includes Columbia Blood Agar Base and Defibulated Horse Blood.

  • Bottom Layer: Columbia BA Base
  • Top Layer: Columbia BA Base and Defibulated Horse Blood
  • Fill Volume: 20 mL
  • Size: 15 mm x 100 mm
  • Prepared Medium

Its fill volume is 20 mL.

NEL manufactures over 500 proprietary products and over 1300 classic media formulations for clients in the pharmaceutical, hospital, food and beverage, cosmetic, and educational research industries.

 : ISO Class 5 / FS209E Class 100 Validated EC Rooms certified.