Order Entry
Puerto Rico
ContactUsLinkComponent
Endo Agar, Plates, Northeast Lab Services
Endo Agar, Plates, Northeast Lab Services
Catalog # 101373-980
CAS Number:  
undefined
Endo Agar, Plates, Northeast Lab Services
Catalog # 101373-980
Supplier Number:  P1305
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Media Format:
    Liquid
  • Description:
    Endo Agar
  • Size:
    15 mm x 100 mm
  • Cat. No.:
    101373-980
  • Fill volume:
    20 ml
  • Supplier no.:
    P1305

Specifications

About this item

Endo Agar is contained in a standard petri dish.

  • Standard Petri Dish
  • Fill Volume: 20 mL
  • Size: 15 mm x 100 mm
  • Prepared Medium

The agar is contained in a standard petri dish measuring 15 mm x 100 mm

NEL's plated media operation utilizes carefully selected and validated ingredients for contaminant-free products.

Certifications: ISO Class 5 / FS209E Class 100 Validated EC Rooms certified.