Order Entry
Puerto Rico
ContactUsLinkComponent
Columbia CNA Agar, Plates, Northeast Lab Services
Columbia CNA Agar, Plates, Northeast Lab Services
Catalog # 101373-956
CAS Number:  
undefined
Columbia CNA Agar, Plates, Northeast Lab Services
Catalog # 101373-956
Supplier Number:  P1250
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Media Format:
    Liquid
  • Description:
    Columbia CNA Agar with Five Percent Sheep's Blood
  • Size:
    15 mm x 100 mm
  • Cat. No.:
    101373-956
  • Fill volume:
    20 ml
  • Supplier no.:
    P1250

Specifications

About this item

Columbia CNA Agar is a prepared medium that contains five percent sheep's blood.

  • Five Percent Sheep's Blood
  • Standard Petri Dish
  • Fill Volume: 20 mL
  • Size: 15 mm x 100 mm
  • Prepared Medium

NEL produces a wide variety of agar-based media in many sizes and types of bottles, both glass and autoclavable plastic.

NEL manufactures over 500 proprietary products and over 1300 classic media formulations for clients in the pharmaceutical, hospital, food and beverage, cosmetic, and educational research industries.

Certifications: ISO Class 5 / FS209E Class 100 Validated EC Rooms certified.