Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Media Format:Liquid
- Description:Brucella Agar with Five Percent Horse's Blood
- Size:15 x 100 mm
- Cat. No.:101373-906
- Fill volume:20 ml
- Supplier no.:P1106
Brucella Agar's formulation includes five percent horse's blood.
- Five Percent Horse's Blood
- Standard Petri Dish
- Fill Volume: 20 mL
- Size: 15 mm x 100 mm
- Prepared Medium
The prepared medium is contained in a standard petri dish.
NEL's careful operation ensures contaminant-free products with a high degree of consistency from batch to batch.