Order Entry
Puerto Rico
ContactUsLinkComponent
Buffered Charcoal Yeast Extract, Plates, Northeast Lab Services
Buffered Charcoal Yeast Extract, Plates, Northeast Lab Services
Catalog # 101373-870
CAS Number:  
Buffered Charcoal Yeast Extract, Plates, Northeast Lab Services
Catalog # 101373-870
Supplier Number:  P1045
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Media Format:
    Liquid
  • Description:
    Buffered Charcoal Yeast Extract with Four Percent L-Cysteine Agar
  • Size:
    15 mm x 100 mm
  • Cat. No.:
    101373-870
  • Fill volume:
    20 ml
  • Supplier no.:
    P1045

Specifications

About this item

Buffered Charcoal Yeast Extract contains four percent L-Cysteine Agar.

  • Four Percent L-Cysteine Agar
  • Standard Petri Dish
  • Fill Volume: 20 mL
  • Size: 15 mm x 100 mm

The extract has a fill volume of 20 mL.

NEL's plated media operation utilizes carefully selected and validated ingredients.

Certifications: ISO Class 5 / FS209E Class 100 Validated EC Rooms certified.