Order Entry
United States
ContactUsLinkComponent
Brain Heart Infusion Agar Slant, Tubes, Northeast Lab Services
Brain Heart Infusion Agar Slant, Tubes, Northeast Lab Services
Catalog # 101374-510
CAS Number:  
Brain Heart Infusion Agar Slant, Tubes, Northeast Lab Services
Catalog # 101374-510
Supplier Number:  T1150
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Media Format:
    Liquid
  • Description:
    Brain Heart Infusion Agar Slant
  • Size:
    16 mm x 125 mm
  • Environmentally Preferable:
  • Cat. No.:
    101374-510
  • Cap type:
    Screw cap
  • Fill volume:
    8 ml
  • Supplier no.:
    T1150

Specifications

About this item

Brain Heart Infusion Agar Slant includes a durable screw cap and has a fill volume of 8 mL.

  • Screw Cap
  • Fill Volume: 8 mL
  • Tube Size: 16 mm x 125 mm
  • Prepared Medium
  • Glass Construction

The glass tube measures 16 mm x 125 mm.

NEL produces slants and other media tubes for a wide variety of clients including private laboratories, food processors, clinics and hospitals, academic institutions, and government agencies.