Order Entry
Puerto Rico
ContactUsLinkComponent
Fluid Thioglycollate, Bottles, Northeast Lab Services
Fluid Thioglycollate, Bottles, Northeast Lab Services
Catalog # 101373-706
CAS Number:  
Fluid Thioglycollate, Bottles, Northeast Lab Services
Catalog # 101373-706
Supplier Number:  B4200
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Media Format:
    Liquid
  • Description:
    Fluid Thioglycollate
  • Size:
    4 oz.
  • Cat. No.:
    101373-706
  • Cap type:
    Aluminum crimp cap
  • Fill volume:
    100 ml
  • Supplier no.:
    B4200

Specifications

About this item

Fluid Thioglycollate includes a polycarbonate serum bottle, a rubber stopper, and an aluminum crimp cap.

  • Polycarbonate Serum Bottle
  • Rubber Stopper
  • Aluminum Crimp Cap
  • Fill Volume: 100 mL
  • Bottle Size: 4 oz.

The serum bottle is 100 mL, and Thioglycollate has a fill volume of 100 mL.

NEL manufactures over 500 proprietary products and over 1300 classic media formulations for clients in the pharmaceutical, hospital, food and beverage, cosmetic, and educational research industries.

Certifications: Follows FDA, GMP, and USDA guidelines.
Delivery: Prepared media in an unsealed container.