Dichloran Agar, Plates, Northeast Lab Services
Catalog # 101373-968
Supplier: Northeast Lab Services
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Media Format:Liquid
- Description:Dichloran Glycerol Agar (DG18)
- Size:15 mm x 100 mm
- Cat. No.:101373-968
- Fill volume:20 ml
- Volume:20 ml
- Supplier no.:P1295
Specifications
About this item
Dichloran Agar is available in several media formulations.
- Standard Petri Dish
- Prepared Medium
- Dichloran Glycerol and DRBC Agar Available
- Plate Size: 15 mm x 100 mm
All media formulations of Dichloran Agar are contained in a 15 x 100 mm petri dish.
NEL produces a wide variety of agar-based media.
Certifications: ISO Class 5 / FS209E Class 100 Validated EC Rooms certified.