Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Media Format:Liquid
- Description:Brain Heart Infusion Agar
- Size:15 mm x 100 mm
- Cat. No.:101373-930
- Fill volume:20 ml
- Supplier no.:P1122
Specifications
About this item
Brain Heart Infusion Agar is a prepared medium contained in a standard petri dish.
- Standard Petri Dish
- Prepared Medium
- Tetracycline
- Size: 15 mm x 100 mm
- Fill Volume: 20 mL
The agar has a fill volume of 20 mL.
NEL manufactures products and media formulations for clients in the pharmaceutical, hospital, food and beverage, cosmetic, and educational research industries.