About this item
MacConkey Agar is available with or without sorbitol.
- Sorbitol Available
- Prepared Medium
- Fill Volume: 20 mL
- Size: 15 mm x 100 mm
- Standard Petri Dish
Both formations are contained in a standard petri dish.
NEL manufactures over 500 proprietary products and over 1300 classic media formulations for clients in the pharmaceutical, hospital, food and beverage, cosmetic, and educational research industries.