About this item
Cooked Meat features different media formulation options.
- Screw Cap
- Glucose, Hemin, and Vitamin K Available
- Medium Cooked Meet Available
- Fill Volume: 10 mL
- Size: 16 mm x 125 mm
From simple fermentation tubes to highly specialized clinical diagnostic media, NEL produces media tubes for a wide variety of clients including private laboratories, food processors, clinics and hospitals, academic institutions, and government agencies.