ContactUsLinkComponent
EE Broth, Bottles, Northeast Lab Services
EE Broth, Bottles, Northeast Lab Services
  101373-404
 :  
EE Broth, Bottles, Northeast Lab Services
  101373-404
 :  B1261
 :  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Media Format:
    Liquid
  • Description:
    EE Broth (Mossel)
  • Size:
    1 L
  • Cat. No.:
    101373-404
  • Cap type:
    33/430 black PP cap
  • Fill volume:
    900 ml
  • Supplier no.:
    B1261
Clear, one-liter EE Broth is available in a variety of size for all food-safety maintenance needs.
  • Polycarbonate Construction
  • Polypro Caps
  • Clear Bottles
  • One Liter
A polycarbonate bottle with a sturdy polypro cap contains the broth.
NEL manufactures over 500 proprietary products and over 1300 classic media formulations for clients in the pharmaceutical, hospital, food and beverage, cosmetic, and educational research industries.
 : Follows FDA, GMP, and USDA guidelines.