Order Entry
Puerto Rico
ContactUsLinkComponent
Bovine Fibrinogen (from Plasma), MP Biomedicals
Bovine Fibrinogen (from Plasma), MP Biomedicals
Supplier:  MP Biomedicals
Certificates
undefined
Bovine Fibrinogen (from Plasma), MP Biomedicals
Supplier:  MP Biomedicals
Certificates

Specifications

  • Source:
    Plasma
  • Species:
    Bovine
  • Storage Conditions:
    Store product at <-10 °C
  • Form:
    Powder
  • Protein/Peptide Name:
    Fibrinogen
  • Purity:
    Protein: 70% w/w, balance excipients
  • Solubility:
    Soluble in saline. Clear to slightly, hazy solution.
  • Color:
    White
  • pH:
    6.4 (1% w/v in saline )
  • Grade:
    Diagnostic Grade
  • Tested Applications:
    Substrate for clotting assays

Specifications

Related Information

Frequently Bought Together