Specifications
- Light Source:LED
- Description:MD 600 Multiparameter Photometer
- Environmentally Preferable:
- Wavelength Accuracy:±1 nm
- Display:Graphic display
- Filter Type:Interference filters
- Interfaces:IR interface for data transfer, RJ45 connector for internet updates
- Keypad:Acid and solvent resistant touch-sensitive keypad
- Operating conditions:5–40°C; 30–90% RH
- Wavelengths:430, 530, 560, 580, 610, 660 nm
- Weight:0,45 kg
- W×D×H:210×95×45 mm
- Cat. No.:89212-658
- Supplier no.:MC500-10
Specifications
About this item
The MD 500 is a fully portable 6 LED, multiparameter photometer. MD 600 is a completely functional integrated unit designed for the outdoor professional for on-site visits.
- Contains more than 120 programmed methods
- Waterproof, easy to handle
- Automatic wavelength selection
- Uses tablet, liquids, powders or vials
- Storage of up to 1000 data sets
The modern design of the MD 500 succeeds in combining the mobility of a portable photometer with the characteristics of a modern laboratory photometer
The MD 500 can also be used with the optional infrared data transfer module which is supplied complete with data logging software, providing additional data manipulation tools and allowing the data to be saved as a spreadsheet or text file.
This unit covers all the important parameters of water analysis, from aluminium to zinc. The high level of accuracy of Lovibond® reagents and the user-friendly nature of the instrument guarantee rapid and reliable analysis.