Accessories for Reagent Reservoirs, Disposable, Sorenson Bioscience
Catalog # 89232-346
Supplier: Sorenson Bioscience
Specifications
- Color:Black
- Description:Reservoir Holder, Open Basin, SBS Footprint
- Packed:Individually wrapped
- Cat. No.:89232-346
- Supplier no.:74020
Specifications
About this item
Re-usable Reservoir Holder is on an SBS Footprint and is designed to lock reservoirs steadily and securely in place for use with the BenchTop Pipettor.