Orders LinkContactUsLinkComponent
Vacuum Microanalysis Holders, Advantec MFS
Vacuum Microanalysis Holders, Advantec MFS
Marketsource
Supplier:   Advantec MFS
Vacuum Microanalysis Holders, Advantec MFS
Supplier:  Advantec MFS
Marketsource
Certificates
4 Options Available Below
Product Details & Documents
13 mm Glass Microanalysis Holders.
  • Concentrate samples in a small area (1.2 cm²)
  • KG 13 AA features a two-part funnel (13 mL/100 mL)
  • Filters small volume of liquid
Specifications for Product Family
Specifications
  • Catalog No:
  • 101104-768
  • 101105-174
  • 101105-684
  • 101104-780
  • Filter Size:
  • 25 mm
  • 25 mm
  • 25 mm
  • 25 mm
Specifications
Product Family Options
Product Information
  • Capacity
    Availability
    Price
  • KG 13 B
    500 ml
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $441.75
    $441.75
    Pack of 1
       $441.75
    Minimum order quantity is 1
    Supplier Number: 311050
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    101104-768
    Filter Size
    25 mm
    Marketsource
  • KG 13 A
    100 ml
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $346.82
    $346.82
    Pack of 1
       $346.82
    Minimum order quantity is 1
    Supplier Number: 311000
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    101105-174
    Filter Size
    25 mm
    Marketsource
  • 101105-684
    KG 13 AA
    KG 13 AA
    13/100 ml
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $459.45
    $459.45
    Pack of 1
       $459.45
    Minimum order quantity is 1
    Supplier Number: 311100
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    101105-684
    Filter Size
    25 mm
    Marketsource
  • KG 13 C
    1000 ml
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $474.49
    $474.49
    Pack of 1
       $474.49
    Minimum order quantity is 1
    Supplier Number: 311110
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    101104-780
    Filter Size
    25 mm
    Marketsource
Accessories
Product Information
  • Catalog #
    Details
    Fits
    Availability
    Price
  • Silicone rubber stopper, #8b
    Silicone rubber stopper, #8b
    101103-602
    -
    KP 47
    Log in for availability
    Pack of 1 - $27.17
Product Information
  • Catalog #
    Availability
    Price
  • Base (Glass frit)
    Base (Glass frit)
    101103-694
    Log in for availability
    Pack of 1 - $309.22
Product Information
  • Catalog #
    Availability
    Price
  • Funnel, 1000 mL
    Funnel, 1000 mL
    101105-862
    Log in for availability
    Pack of 1 - $485.08