ContactUsLinkComponent
IsoGel® Isoelectric-Focusing Agarose, Lonza
  12001-794
 :  Lonza
 :  
undefined
IsoGel® Isoelectric-Focusing Agarose, Lonza
  12001-794
 :  Lonza
 :  50202
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    IsoGel® Isoelectric-Focusing Agarose
  • Size:
    25 g
  • Environmentally Preferable:
  • Cat. No.:
    12001-794
  • Application:
    Isoelectic focusing, IEF applications
  • Supplier no.:
    50202
IsoGel® Agarose is designed specifically for IEF (Isoelectric focusing) applications
  • Designed for IEF
  • Rigid enough for vertical casting
  • Low EEO
  • Transparent for record keeping
IsoGel(R) Agarose is a highly purified agarose specifically manufactured for isoelectric focusing (IEF). It has no measurable EEO due to a manufacturing process which minimizes fixed anions and mobile cations. IsoGel(R) Agarose IEF Plates are sufficiently rigid to be cast in vertical tubes in addition to vertically molded and horizontally open cast thin gels. Gels cast on GelBond® Film dry down quickly and easily to transparent permanent records.