About this item
Interleukin 17E (IL-17E), also known as IL-25, is a member of the IL-17 family of cytokines. IL-17E binds to the IL-17RB receptor to stimulate the secretion of the proinflammatory interleukin 8 (IL-8) chemokine and to induce the activation of nuclear factor kappa-light-chain-enhancer of activated B cells (NF-κB).
- High quality
- Low endotoxin
- Affordable
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage Conditions:–20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Biological Activity:Bioactivity: No biological activity data is available at this time
- Gene ID:Q8VHC9
- Reconstitution Instructions:Sterile 10 mM HCl at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:Y
- Protein Synonyms:IL-25
- Protein/Peptide Name:IL-17E (AF)
- Purity:≥90%
- Molecular Weight:17.6/35.2 kDa
- Sequence:MVSLRIQEGC SHLPSCCPSK EQEPPEEWLK WSSASVSPPE PLSHTHHAES CRASKDGPLN SRAISPWSYE LDRDLNRVPQ DLYHARCLCP HCVSLQTGSH MDPLGNSVPL YHNQTVFYRR PCHGEEGTHR RYCLERRLYR VSLACVCVRP RVMA
- Endotoxin Level:Low
- Formulation:0.1% Trifluoroacetic acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT