BioClean™ MEA™ Sterile Looped Facemask, Ansell
Catalog # 75836-208
Supplier: Ansell Healthcare
Specifications
- Sterility:Sterile
- Material:PE
- Description:BioClean MEA™ sterile looped facemask
- Cat. No.:75836-208
- Supplier no.:MEA210-1
Specifications
About this item
BioClean MEA™ facemask offers ultra-low particulation.
- High strength seams
- 3-ply design with ultrasonic sealing
- High BFE and PFE
- Loops with blue plastic clip connector allow secure fastening at back of head
- Outer Facing Layer: Non-woven spunbonded PE (hydrophobic)
Providing high particle and bacterial filtration efficiency, the latex-free and low-linting BioClean MEA™ sterile looped facemask combines reliable protection with comfort and exceptional breathability.
Certifications: ISO Class 4 and EU GMP Grade A compatible
Manufactured in a facility holding ISO 9001:2008.
Processed in an NEBB certified ISO Class 4 cleanroom environment.
Sampling and inspection in accordance with ISO 2859-3:1994.
Tested against standard ASTM F2101 for Bacterial Filtration Efficiency (BFE).
Manufactured in a facility holding ISO 9001:2008.
Processed in an NEBB certified ISO Class 4 cleanroom environment.
Sampling and inspection in accordance with ISO 2859-3:1994.
Tested against standard ASTM F2101 for Bacterial Filtration Efficiency (BFE).
Packaging: Face masks are packaged one piece (with blue clip) per sealed inner PE bag; 50 inner bags per sealed outer PE bag; 12 outer bags per lined carton (600 pieces).